missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl Itaconate (stabilized with TBC) 98.0+%, TCI America™
$143.54 - $1168.76
Chemical Identifiers
| CAS | 2409-52-1 |
|---|---|
| Molecular Formula | C9H14O4 |
| Molecular Weight (g/mol) | 186.207 |
| MDL Number | MFCD00059230 |
| InChI Key | ZEFVHSWKYCYFFL-UHFFFAOYSA-N |
| Synonym | Itaconic Acid Diethyl Ester |
| PubChem CID | 75481 |
| IUPAC Name | diethyl 2-methylidenebutanedioate |
| SMILES | CCOC(=O)CC(=C)C(=O)OCC |
Chemical Identifiers
| 2409-52-1 | |
| 186.207 | |
| ZEFVHSWKYCYFFL-UHFFFAOYSA-N | |
| 75481 | |
| CCOC(=O)CC(=C)C(=O)OCC |
| C9H14O4 | |
| MFCD00059230 | |
| Itaconic Acid Diethyl Ester | |
| diethyl 2-methylidenebutanedioate |
Specifications
| 2409-52-1 | |
| 228°C | |
| MFCD00059230 | |
| Itaconic Acid Diethyl Ester | |
| CCOC(=O)CC(=C)C(=O)OCC | |
| 186.207 | |
| 186.21 | |
| Liquid |
| Yellow | |
| C9H14O4 | |
| 25 g | |
| ZEFVHSWKYCYFFL-UHFFFAOYSA-N | |
| diethyl 2-methylidenebutanedioate | |
| 75481 | |
| ≥98.0% (GC) | |
| Diethyl Itaconate (stabilized with TBC) |
Safety and Handling
EINECSNumber : (2)-1130&(2)-1142
TSCA : Yes