missing translation for 'onlineSavingsMsg'
Learn More

Diethyl ethylidenemalonate, 99%

CAS: 1462-12-0 | C9H14O4 | 186.21 g/mol

$50.48 - $130.99

Chemical Identifiers

CAS 1462-12-0
Molecular Formula C9H14O4
Molecular Weight (g/mol) 186.21
MDL Number MFCD00009145
InChI Key LBBAWVLUOZVYCC-UHFFFAOYSA-N
Synonym diethyl ethylidenemalonate, diethyl 2-ethylidenemalonate, propanedioic acid, ethylidene-, diethyl ester, 1,3-diethyl 2-ethylidenepropanedioate, 2-ethylidenemalonic acid diethyl ester, diethylethylidenemalonate, acmc-1c0mi, diethyl ethylidene malonate, diethyl ethylidenepropanedioate, diethyl 2-ethylidenemalonate #
PubChem CID 73831
SMILES CCOC(=O)C(=CC)C(=O)OCC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1211703
View Documents
Thermo Scientific Chemicals
L1211703
1 g
Each for $50.48
Only null left
 
AAL1211706
View Documents
Thermo Scientific Chemicals
L1211706
5 g
Each for $130.99
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1462-12-0
186.21
LBBAWVLUOZVYCC-UHFFFAOYSA-N
73831
C9H14O4
MFCD00009145
diethyl ethylidenemalonate, diethyl 2-ethylidenemalonate, propanedioic acid, ethylidene-, diethyl ester, 1,3-diethyl 2-ethylidenepropanedioate, 2-ethylidenemalonic acid diethyl ester, diethylethylidenemalonate, acmc-1c0mi, diethyl ethylidene malonate, diethyl ethylidenepropanedioate, diethyl 2-ethylidenemalonate #
CCOC(=O)C(=CC)C(=O)OCC

Specifications

1462-12-0
115°C to 116°C (17 mmHg)
C9H14O4
MFCD00009145
1773932
LBBAWVLUOZVYCC-UHFFFAOYSA-N
diethyl 2-ethylidenepropanedioate
73831
99%
1.019
>110°C (230°F)
1.442
1 g
diethyl ethylidenemalonate, diethyl 2-ethylidenemalonate, propanedioic acid, ethylidene-, diethyl ester, 1,3-diethyl 2-ethylidenepropanedioate, 2-ethylidenemalonic acid diethyl ester, diethylethylidenemalonate, acmc-1c0mi, diethyl ethylidene malonate, diethyl ethylidenepropanedioate, diethyl 2-ethylidenemalonate #
CCOC(=O)C(=CC)C(=O)OCC
186.21
186.21
Diethyl ethylidenemalonate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 215-965-5

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only