missing translation for 'onlineSavingsMsg'
Learn More

Diethyl (Ethoxymethyl)phosphonate 98.0+%, TCI America™

Catalog No. p-7128653
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D50955G 5 g
D509525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D50955G Supplier TCI America Supplier No. D50955G
Only null left

Chemical Identifiers

CAS 10419-80-4
Molecular Formula C7H17O4P
Molecular Weight (g/mol) 196.18
MDL Number MFCD00137939
InChI Key PELDURRTQRGRMV-UHFFFAOYSA-N
Synonym (Ethoxymethyl)phosphonic Acid Diethyl Ester
PubChem CID 13128691
IUPAC Name diethyl (ethoxymethyl)phosphonate
SMILES CCOCP(=O)(OCC)OCC

Specifications

Color Colorless
Boiling Point 103°C
Quantity 5 g
Formula Weight 196.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl (Ethoxymethyl)phosphonate