accessibility menu, dialog, popup

UserName

CH3OPOOH3COH3C

CAS RN 10419-80-4

IUPAC Name: diethyl (ethoxymethyl)phosphonate
Synonyms: diethyl ethoxymethylphosphonate
Molecular Weight (g/mol): 196.18
Molecular Formula: C7H17O4P
InChi Key: PELDURRTQRGRMV-UHFFFAOYSA-N
SMILES: CCOCP(=O)(OCC)OCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results