missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl Ether, puriss., ≥99.5% (GC), Honeywell™ Riedel-de Haën™
Puriss., meets analytical specification of BP, Ph. Eur.,≥99.5% (GC)
Supplier: Honeywell-Riedel-de Haen 240044X25L
Description
- Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
- It is the leading brand offering High-Purity Solvents for dedicated applications.
Specifications
| Diethyl Ether | |
| Meets analytical specification of Ph.Eur.BP | |
| 0.706g/mL at 25°C | |
| 4 x 2.5 L | |
| −45°C (−49°F) | |
| Glass Bottle | |
| C4H10O | |
| MFCD00011646 | |
| 1696894 | |
| RTZKZFJDLAIYFH-UHFFFAOYSA-N | |
| ethoxyethane | |
| 3283 | |
| 74.12g/mol | |
| puriss. |
| -116°C | |
| 60-29-7 | |
| 34.6°C | |
| 0.365 | |
| 320°F | |
| Liquid | |
| (CH3CH2)2O | |
| UN1155 | |
| diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether | |
| CCOCC | |
| 74.12 | |
| CHEBI:35702 | |
| ≥99.5% (GC) |
Chemical Identifiers
| 60-29-7 | |
| 74.12 | |
| RTZKZFJDLAIYFH-UHFFFAOYSA-N | |
| 3283 | |
| ethoxyethane |
| C4H10O | |
| MFCD00011646 | |
| diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether | |
| CHEBI:35702 | |
| CCOCC |
Safety and Handling
P210-P261
H224-H302-H336
DOTInformation : class 3 - PG 1 - Diethyl ether
EINECSNumber : 200-467-2
RTECSNumber : KI5775000