missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Ether, puriss., ≥99.5% (GC), Honeywell™ Riedel-de Haën™

Catalog No. p-7120380
Change view
Click to view available options
Quantity:
1 L
2.5 L
5 L
4 x 2.5 L
4 x 5 L
6 x 1 L
135 kg
Packaging:
Aluminum Bottle
Glass Bottle
Metal Drum
Catalog No. Quantity Packaging
60045503 1 L Glass Bottle
60045504 2.5 L Glass Bottle
60045510 5 L Aluminum Bottle
60045512 6 x 1 L Glass Bottle
60045508 4 x 2.5 L Glass Bottle
60045509 4 x 5 L Aluminum Bottle
60045502 135 kg Metal Drum
7 options

Catalog No. 60045503

Supplier: Honeywell Inc 240041L

Only null left

Puriss., meets analytical specification of BP, Ph. Eur.,≥99.5% (GC)

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.

Chemical Identifiers

CAS 60-29-7
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
MDL Number MFCD00011646
InChI Key RTZKZFJDLAIYFH-UHFFFAOYSA-N
Synonym diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
PubChem CID 3283
ChEBI CHEBI:35702
IUPAC Name ethoxyethane
SMILES CCOCC

Specifications

Melting Point -116°C
Density 0.706g/mL at 25°C
Boiling Point 34.6°C
Quantity 1 L
Explosion Limit 0.365
Flash Point −45°C (−49°F)
Ignition Point 320°F
Packaging Glass Bottle
Physical Form Liquid
Linear Formula (CH3CH2)2O
UN Number UN1155
Beilstein 1696894
Formula Weight 74.12g/mol
Percent Purity ≥99.5% (GC)
Grade puriss.
Chemical Name or Material Diethyl Ether, Meets analytical specification of Ph.Eur.BP
Show More Show Less
Hazard Statement P210-P261
Precautionary Statement H224-H302-H336
DOTInformation class 3 - PG 1 - Diethyl ether
EINECSNumber 200-467-2
RTECSNumber KI5775000