missing translation for 'onlineSavingsMsg'
Learn More

Diethyl ether, ≥99.9% (GC), MilliporeSigma™ Supelco™

Catalog No. p-7187572
Change view
Click to view available options
Quantity:
1 mL
10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020382 1 mL
111018347 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111020382 Supplier Merck Emd Millipore Supplier No. 912381MLF
Only null left

Diethyl ether is a colorless and highly volatile organic compound, which commonly finds applications as an extraction solvent.

Featured Industries: Environmental

Specifications

CAS 60-29-7
Ignition Point 320°F
Solution Contains ˜0.0005% 2,6-di- tert. -butyl-4-methylphenol as stabilizer
Vapor Pressure 2311 hPa (60°C)
Vapor Density 2.6 (vs. air)
Density 0.706 g/mL (at 25°C)
Flash Point -40°C
Physical Form Neat
Linear Formula (CH3 CH2)2 O
MDL Number MFCD00011646
Melting Point -116°C
Boiling Point 34.6°C
Synonym Ether; Ethyl ether
Type Analytical Standard
Quantity 1 mL
Formula Weight 74.12
Percent Purity ≥99.9% (GC)
Grade Analytical Standard
Show More Show Less