missing translation for 'onlineSavingsMsg'
Learn More

Diethyl DL-Malate 98.0+%, TCI America™

Supplier:  TCI America M0024500G

 View more versions of this product

Catalog No. M0024500G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Diethyl DL-Malate
Yellow
C8H14O5
500 g
VKNUORWMCINMRB-UHFFFAOYNA-N
diethyl 2-hydroxybutanedioate
24197
190.20
Liquid
7554-12-3
255°C
MFCD00048556
DL-Apple Acid Diethyl Ester, DL-Malic Acid Diethyl Ester
CCOC(=O)CC(C(=O)OCC)O
190.20
CHEBI:87368
≥98.0% (GC)

Chemical Identifiers

7554-12-3
190.20
VKNUORWMCINMRB-UHFFFAOYNA-N
24197
diethyl 2-hydroxybutanedioate
C8H14O5
MFCD00048556
DL-Apple Acid Diethyl Ester, DL-Malic Acid Diethyl Ester
CHEBI:87368
CCOC(=O)CC(C(=O)OCC)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1444

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions