missing translation for 'onlineSavingsMsg'
Learn More

Diethyl DL-Malate 98.0+%, TCI America™

Catalog No. M002425G
Change view
Click to view available options
Quantity:
25 g
500 g
Catalog No. Quantity
M002425G 25 g
M0024500G 500 g
2 options

Catalog No. M002425G

Supplier: TCI America M002425G

Only null left

Chemical Identifiers

CAS 7554-12-3
Molecular Formula C8H14O5
Molecular Weight (g/mol) 190.20
MDL Number MFCD00048556
InChI Key VKNUORWMCINMRB-UHFFFAOYNA-N
Synonym DL-Apple Acid Diethyl Ester, DL-Malic Acid Diethyl Ester
PubChem CID 24197
ChEBI CHEBI:87368
IUPAC Name diethyl 2-hydroxybutanedioate
SMILES CCOC(=O)CC(C(=O)OCC)O

Specifications

Color Yellow
Boiling Point 255°C
Quantity 25 g
Formula Weight 190.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl DL-Malate
EINECSNumber (2)-1444
TSCA Yes