missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl Diselenide 96.0+%, TCI America™
$193.86 - $680.38
Chemical Identifiers
| CAS | 628-39-7 |
|---|---|
| Molecular Formula | C4H10Se2 |
| Molecular Weight (g/mol) | 216.066 |
| MDL Number | MFCD00015491 |
| InChI Key | OMAWWKIPXLIPDE-UHFFFAOYSA-N |
| Synonym | Ethyl Diselenide |
| PubChem CID | 69405 |
| IUPAC Name | (ethyldiselanyl)ethane |
| SMILES | CC[Se][Se]CC |
Chemical Identifiers
| 628-39-7 | |
| 216.066 | |
| OMAWWKIPXLIPDE-UHFFFAOYSA-N | |
| 69405 | |
| CC[Se][Se]CC |
| C4H10Se2 | |
| MFCD00015491 | |
| Ethyl Diselenide | |
| (ethyldiselanyl)ethane |
Specifications
| 628-39-7 | |
| C4H10Se2 | |
| 1 g | |
| Ethyl Diselenide | |
| CC[Se][Se]CC | |
| 216.066 | |
| 216.04 | |
| Liquid |
| 85°C | |
| MFCD00015491 | |
| 3440 | |
| OMAWWKIPXLIPDE-UHFFFAOYSA-N | |
| (ethyldiselanyl)ethane | |
| 69405 | |
| ≥96.0% (GC) | |
| Diethyl Diselenide |
Safety and Handling
RTECSNumber : JM9151750
TSCA : Yes
Recommended Storage : Refrigerator