accessibility menu, dialog, popup

UserName

H3CSeSeCH3

CAS RN 628-39-7

IUPAC Name: (ethyldiselanyl)ethane
Synonyms: diselenide, diethyl
Molecular Weight (g/mol): 216.07
Molecular Formula: C4H10Se2
InChi Key: OMAWWKIPXLIPDE-UHFFFAOYSA-N
SMILES: CC[Se][Se]CC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results