missing translation for 'onlineSavingsMsg'
Learn More

Diethyl D-(-)-Tartrate 97.0+%, TCI America™

Catalog No. p-7136651
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T119525G 25 g
T1195250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T119525G Supplier TCI America Supplier No. T119525G
Only null left

Chemical Identifiers

CAS 13811-71-7
Molecular Formula C8H14O6
Molecular Weight (g/mol) 206.19
MDL Number MFCD00064451
InChI Key YSAVZVORKRDODB-WDSKDSINSA-N
Synonym --diethyl d-tartrate, 2s,3s-diethyl 2,3-dihydroxysuccinate, diethyl d-tartrate, d---tartaric acid diethyl ester, diethyl d---tartrate, diethyl-d-tartrate, diethyl 2s,3s-2,3-dihydroxybutanedioate, 2s,3s--dihydroxybutane-1,4-dioic acid diethyl ester, --diethyl-d-tartrate, diethyl s-r*,r*-tartrate
PubChem CID 117410
IUPAC Name 1,4-diethyl (2S,3S)-2,3-dihydroxybutanedioate
SMILES CCOC(=O)[C@@H](O)[C@H](O)C(=O)OCC

Specifications

Color Yellow
Boiling Point 130°C
Quantity 25 g
Formula Weight 206.19
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl D-(-)-Tartrate
EINECSNumber (2)-1458
TSCA Yes