missing translation for 'onlineSavingsMsg'
Learn More

(-)-Diethyl D-tartrate, 99%

CAS: 13811-71-7 | C8H14O6 | 206.19 g/mol

$52.44 - $391.90

Chemical Identifiers

CAS 13811-71-7
Molecular Formula C8H14O6
Molecular Weight (g/mol) 206.19
MDL Number MFCD00064451
InChI Key YSAVZVORKRDODB-WDSKDSINSA-N
Synonym --diethyl d-tartrate, 2s,3s-diethyl 2,3-dihydroxysuccinate, diethyl d-tartrate, d---tartaric acid diethyl ester, diethyl d---tartrate, diethyl-d-tartrate, diethyl 2s,3s-2,3-dihydroxybutanedioate, 2s,3s--dihydroxybutane-1,4-dioic acid diethyl ester, --diethyl-d-tartrate, diethyl s-r*,r*-tartrate
PubChem CID 117410
IUPAC Name diethyl (2S,3S)-2,3-dihydroxybutanedioate
SMILES CCOC(=O)[C@@H](O)[C@H](O)C(=O)OCC
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1799206
View Documents
Thermo Scientific Chemicals
A1799206
5 g N/A
Only null left
 
AAA1799214
View Documents
Thermo Scientific Chemicals
A1799214
25 g N/A
Only null left
 
AAA1799222
View Documents
Thermo Scientific Chemicals
A1799222
100 g N/A
Only null left
 
Description

Description

Diethyl D(-)-tartrate is mainly used in chiral pharmaceuticals, chiral intermediates and chiral catalysts. It can be used with titanium in enolic asymmetric epoxidization. The compound d-alpha-tocopherol is the principal compound present in natural vitamin E sources. At present, d-alpha-tocopherol and other tocopherol derivatives are used in pharmaceuticals, foods and animal feeds. It is used as a food additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl D(-)-tartrate is mainly used in chiral pharmaceuticals, chiral intermediates and chiral catalysts. It can be used with titanium in enolic asymmetric epoxidization. The compound d-alpha-tocopherol is the principal compound present in natural vitamin E sources. At present, d-alpha-tocopherol and other tocopherol derivatives are used in pharmaceuticals, foods and animal feeds. It is used as a food additive.

Solubility
Insoluble in water.

Notes
Air & Moisture Sensitive. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Chemical Identifiers

13811-71-7
206.19
YSAVZVORKRDODB-WDSKDSINSA-N
117410
CCOC(=O)[C@@H](O)[C@H](O)C(=O)OCC
C8H14O6
MFCD00064451
--diethyl d-tartrate, 2s,3s-diethyl 2,3-dihydroxysuccinate, diethyl d-tartrate, d---tartaric acid diethyl ester, diethyl d---tartrate, diethyl-d-tartrate, diethyl 2s,3s-2,3-dihydroxybutanedioate, 2s,3s--dihydroxybutane-1,4-dioic acid diethyl ester, --diethyl-d-tartrate, diethyl s-r*,r*-tartrate
diethyl (2S,3S)-2,3-dihydroxybutanedioate

Specifications

13811-71-7
1.205
93°C (199°F)
1.446
5 g
--diethyl d-tartrate, 2s,3s-diethyl 2,3-dihydroxysuccinate, diethyl d-tartrate, d---tartaric acid diethyl ester, diethyl d---tartrate, diethyl-d-tartrate, diethyl 2s,3s-2,3-dihydroxybutanedioate, 2s,3s--dihydroxybutane-1,4-dioic acid diethyl ester, --diethyl-d-tartrate, diethyl s-r*,r*-tartrate
YSAVZVORKRDODB-WDSKDSINSA-N
CCOC(=O)[C@@H](O)[C@H](O)C(=O)OCC
206.19
206.19
(-)-Diethyl D-tartrate
17°C
280°C
C8H14O6
MFCD00064451
1727143
Insoluble in water.
−26.5° (c=1 in Water)
diethyl (2S,3S)-2,3-dihydroxybutanedioate
117410
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

P210-P280-P370+P378q-P501c

H227

EINECSNumber : 237-458-8

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only