missing translation for 'onlineSavingsMsg'
Learn More

(-)-Diethyl D-tartrate, 99%

Catalog No. AAA1799206
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1799206 5 g
AAA1799214 25 g
AAA1799222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1799206 Supplier Thermo Scientific Chemicals Supplier No. A1799206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 13811-71-7 | C8H14O6 | 206.19 g/mol

Diethyl D(-)-tartrate is mainly used in chiral pharmaceuticals, chiral intermediates and chiral catalysts. It can be used with titanium in enolic asymmetric epoxidization. The compound d-alpha-tocopherol is the principal compound present in natural vitamin E sources. At present, d-alpha-tocopherol and other tocopherol derivatives are used in pharmaceuticals, foods and animal feeds. It is used as a food additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl D(-)-tartrate is mainly used in chiral pharmaceuticals, chiral intermediates and chiral catalysts. It can be used with titanium in enolic asymmetric epoxidization. The compound d-alpha-tocopherol is the principal compound present in natural vitamin E sources. At present, d-alpha-tocopherol and other tocopherol derivatives are used in pharmaceuticals, foods and animal feeds. It is used as a food additive.

Solubility
Insoluble in water.

Notes
Air & Moisture Sensitive. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 13811-71-7
Molecular Formula C8H14O6
Molecular Weight (g/mol) 206.19
MDL Number MFCD00064451
InChI Key YSAVZVORKRDODB-WDSKDSINSA-N
Synonym --diethyl d-tartrate, 2s,3s-diethyl 2,3-dihydroxysuccinate, diethyl d-tartrate, d---tartaric acid diethyl ester, diethyl d---tartrate, diethyl-d-tartrate, diethyl 2s,3s-2,3-dihydroxybutanedioate, 2s,3s--dihydroxybutane-1,4-dioic acid diethyl ester, --diethyl-d-tartrate, diethyl s-r*,r*-tartrate
PubChem CID 117410
IUPAC Name diethyl (2S,3S)-2,3-dihydroxybutanedioate
SMILES CCOC(=O)[C@@H](O)[C@H](O)C(=O)OCC

Specifications

Melting Point 17°C
Density 1.205
Boiling Point 280°C
Flash Point 93°C (199°F)
Refractive Index 1.446
Quantity 5 g
Beilstein 1727143
Solubility Information Insoluble in water.
Optical Rotation −26.5° (c=1 in Water)
Formula Weight 206.19
Percent Purity 99%
Chemical Name or Material (-)-Diethyl D-tartrate
Show More Show Less
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 237-458-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only