missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl D-(+)-Malate 98.0+%, TCI America™
$231.14 - $691.57
Chemical Identifiers
| CAS | 7554-28-1 |
|---|---|
| Molecular Formula | C8H14O5 |
| Molecular Weight (g/mol) | 190.20 |
| MDL Number | MFCD02093483 |
| InChI Key | VKNUORWMCINMRB-UHFFFAOYNA-N |
| Synonym | D-(+)-Apple Acid Diethyl Ester, D-(+)-Malic Acid Diethyl Ester |
| PubChem CID | 1715086 |
| IUPAC Name | 1,4-diethyl 2-hydroxybutanedioate |
| SMILES | CCOC(=O)CC(O)C(=O)OCC |
Chemical Identifiers
| 7554-28-1 | |
| 190.20 | |
| VKNUORWMCINMRB-UHFFFAOYNA-N | |
| 1715086 | |
| CCOC(=O)CC(O)C(=O)OCC |
| C8H14O5 | |
| MFCD02093483 | |
| D-(+)-Apple Acid Diethyl Ester, D-(+)-Malic Acid Diethyl Ester | |
| 1,4-diethyl 2-hydroxybutanedioate |
Specifications
| 7554-28-1 | |
| C8H14O5 | |
| 5 g | |
| VKNUORWMCINMRB-UHFFFAOYNA-N | |
| 1,4-diethyl 2-hydroxybutanedioate | |
| 1715086 | |
| ≥98.0% (GC) | |
| Diethyl D-(+)-Malate |
| Colorless | |
| MFCD02093483 | |
| D-(+)-Apple Acid Diethyl Ester, D-(+)-Malic Acid Diethyl Ester | |
| CCOC(=O)CC(O)C(=O)OCC | |
| 190.20 | |
| 190.20 | |
| Liquid |
Safety and Handling
TSCA : No