CAS RN 7554-28-1
CAS RN 7554-28-1
IUPAC Name:
1,4-diethyl 2-hydroxybutanedioate
Synonyms:
diethyl malate
Molecular Weight (g/mol):
190.2
Molecular Formula:
C8H14O5
InChi Key:
VKNUORWMCINMRB-UHFFFAOYNA-N
SMILES:
CCOC(=O)CC(O)C(=O)OCC
1
–
1
of
1
results
1
Diethyl D-(+)-Malate 98.0+%, TCI America™
CAS: 7554-28-1 Molecular Formula: C8H14O5 Molecular Weight (g/mol): 190.20 MDL Number: MFCD02093483 InChI Key: VKNUORWMCINMRB-UHFFFAOYNA-N Synonym: D-(+)-Apple Acid Diethyl Ester, D-(+)-Malic Acid Diethyl Ester PubChem CID: 1715086 IUPAC Name: 1,4-diethyl 2-hydroxybutanedioate SMILES: CCOC(=O)CC(O)C(=O)OCC