missing translation for 'onlineSavingsMsg'
Learn More

Diethyl D-(+)-Malate 98.0+%, TCI America™

Numéro de catalogue. M13515G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
M13515G 5 g
M135125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. M13515G Fournisseur TCI America Code fournisseur M13515G
Il en reste null

Identifiants chimiques

CAS 7554-28-1
Molecular Formula C8H14O5
Molecular Weight (g/mol) 190.20
MDL Number MFCD02093483
InChI Key VKNUORWMCINMRB-UHFFFAOYNA-N
Synonym D-(+)-Apple Acid Diethyl Ester, D-(+)-Malic Acid Diethyl Ester
PubChem CID 1715086
IUPAC Name 1,4-diethyl 2-hydroxybutanedioate
SMILES CCOC(=O)CC(O)C(=O)OCC

Spécifications

Color Colorless
Quantity 5 g
Formula Weight 190.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl D-(+)-Malate