missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Cyanophosphonate 95.0+%, TCI America™

Catalog No. p-7124872
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C12425G 5 g
C124225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C12425G Supplier TCI America Supplier No. C12425G
Only null left

Chemical Identifiers

CAS 2942-58-7
Molecular Formula C5H10NO3P
Molecular Weight (g/mol) 163.11
MDL Number MFCD00010256
InChI Key ZWWWLCMDTZFSOO-UHFFFAOYSA-N
Synonym diethyl cyanophosphonate, diethyl phosphorocyanidate, diethylphosphorocyanidate, diethylcyanophosphate, diethyl cyanidophosphate, phosphorocyanidic acid, diethyl ester, diethoxy-phosphoryl cyanide, diethyl cyanophosphosphonate, cyanophosphonic acid diethyl ester, diethylcyanophosphonate
PubChem CID 76257
IUPAC Name diethyl cyanophosphonate
SMILES CCOP(=O)(OCC)C#N

Specifications

Color Yellow
Boiling Point 104°C
Quantity 5 g
UN Number 2922
Formula Weight 163.11
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl Cyanophosphonate
EINECSNumber (9)-0563
RTECSNumber TD2500000
TSCA No