missing translation for 'onlineSavingsMsg'
Learn More

Diethyl chlorophosphite, 97%, Thermo Scientific Chemicals

Catalog No. p-7045242
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0991906 5 g
AAL0991914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0991906 Supplier Thermo Scientific Chemicals Supplier No. L0991906
Only null left

Diethyl chlorophosphite acts as a versatile phosphorylating agent for ester and lactone enolates, amines and alkenols. It is used in the synthesis of organophosphates and are considered anticholinesterase agents through observation in studies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl chlorophosphite acts as a versatile phosphorylating agent for ester and lactone enolates, amines and alkenols. It is used in the synthesis of organophosphates and are considered anticholinesterase agents through observation in studies.

Solubility
Soluble in DMSO and chloroform. Reacts with water.

Notes
Air & Moisture Sensitive. Store away from air and strong bases. Incompatible with water, bases, active metals and oxidizing agents.

Chemical Identifiers

CAS 589-57-1
Molecular Formula C4H10ClO2P
Molecular Weight (g/mol) 156.546
MDL Number MFCD00009074
InChI Key TXHWYSOQHNMOOU-UHFFFAOYSA-N
Synonym diethyl chlorophosphite, ethyl phosphorochloridite, diethyl phosphorochloridite, chlorodiethoxyphosphine, phosphorochloridous acid, diethyl ester, unii-f414s3v8k6, diethyl chlorophosphonite, chloro diethoxy phosphane, diethychlorophosphite, ethyl chlorophosphite
PubChem CID 68530
IUPAC Name chloro(diethoxy)phosphane
SMILES CCOP(OCC)Cl

Specifications

Density 1.082
Boiling Point 153°C to 155°C
Flash Point 25°C (77°F)
Odor Acrid
Refractive Index 1.4375
Quantity 5 g
UN Number UN2920
Beilstein 1098392
Sensitivity Air and moisture sensitive
Merck Index 14,3841
Solubility Information Soluble in DMSO and chloroform. Reacts with water.
Formula Weight 156.55
Percent Purity 97%
Chemical Name or Material Diethyl chlorophosphite
Show More Show Less
Hazard Category H226-H314-H335
Hazard Statement GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.
EINECSNumber 209-652-2
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only