missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Chlorooxalacetate 93.0+%, TCI America™

Catalog No. D36351G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D36351G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D36351G Supplier TCI America Supplier No. D36351G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 34034-87-2
Molecular Formula C8H11ClO5
Molecular Weight (g/mol) 222.62
MDL Number MFCD04114359
InChI Key JNQWFLVFHCCWPV-UHFFFAOYNA-N
Synonym Chlorooxalacetic Acid Diethyl Ester
PubChem CID 10998635
IUPAC Name 1,4-diethyl 2-chloro-3-oxobutanedioate
SMILES CCOC(=O)C(Cl)C(=O)C(=O)OCC

Specifications

Color Yellow
Boiling Point 130°C
Quantity 1 g
Formula Weight 222.62
Percent Purity ≥93.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl Chlorooxalacetate
TSCA No
Recommended Storage Refrigerator