accessibility menu, dialog, popup

UserName

CAS RN 34034-87-2

IUPAC Name: 1,4-diethyl 2-chloro-3-oxobutanedioate
Synonyms: 1,4-diethyl 2-chloro-3-oxobutanedioate
Molecular Weight (g/mol): 222.62
Molecular Formula: C8H11ClO5
InChi Key: JNQWFLVFHCCWPV-UHFFFAOYNA-N
SMILES: CCOC(=O)C(Cl)C(=O)C(=O)OCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results