missing translation for 'onlineSavingsMsg'
Learn More

Diethyl chloromalonate, 90%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 114230050

 View more versions of this product

Catalog No. AC114230050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Diethyl chloromalonate
1.2040g/mL
Authentic
C7H11ClO4
1.431
1.204
WLWCQKMQYZFTDR-UHFFFAOYSA-N
diethyl 2-chloropropanedioate
84182
90%
14064-10-9
113°C
88% min. (GC)
C2H5O2CCH(Cl)CO2C2H5
5mL
diethyl chloromalonate, diethyl 2-chloromalonate, ethyl chloromalonate, ethylmonochloromalonate, propanedioic acid, chloro-, diethyl ester, chloromalonic acid, diethyl ester, diethyl chloropropanedioate, malonic acid, chloro-, diethyl ester, 1,3-diethyl 2-chloropropanedioate, propanedioic acid, 2-chloro-, 1,3-diethyl ester
CCOC(=O)C(C(=O)OCC)Cl
194.61
194.61

Chemical Identifiers

14064-10-9
194.61
diethyl chloromalonate, diethyl 2-chloromalonate, ethyl chloromalonate, ethylmonochloromalonate, propanedioic acid, chloro-, diethyl ester, chloromalonic acid, diethyl ester, diethyl chloropropanedioate, malonic acid, chloro-, diethyl ester, 1,3-diethyl 2-chloropropanedioate, propanedioic acid, 2-chloro-, 1,3-diethyl ester
diethyl 2-chloropropanedioate
C7H11ClO4
WLWCQKMQYZFTDR-UHFFFAOYSA-N
84182
CCOC(=O)C(C(=O)OCC)Cl
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Very toxic to aquatic life.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

EINECSNumber : 237-913-

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only