missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Butylmalonate 99.0+%, TCI America™

Catalog No. p-7118412
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B038125ML 25 mL
B0381500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B038125ML Supplier TCI America Supplier No. B038125ML
Only null left

Chemical Identifiers

CAS 133-08-4
Molecular Formula C11H20O4
Molecular Weight (g/mol) 216.28
MDL Number MFCD00009170
InChI Key RPNFNBGRHCUORR-UHFFFAOYSA-N
Synonym diethyl butylmalonate, diethyl n-butylmalonate, butylmalonic acid diethyl ester, diethyl 2-butylmalonate, ethyl butylmalonate, propanedioic acid, butyl-, diethyl ester, unii-cyy2q744wb, cyy2q744wb, n-butylmalonic acid diethyl ester, malonic acid, butyl-, diethyl ester
PubChem CID 67244
IUPAC Name 1,3-diethyl 2-butylpropanedioate
SMILES CCCCC(C(=O)OCC)C(=O)OCC

Specifications

Color Colorless
Boiling Point 240°C
Quantity 25 mL
Formula Weight 216.28
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl Butylmalonate
EINECSNumber (2)-0879
TSCA No