missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl (Bromodifluoromethyl)phosphonate 97.0+%, TCI America™
Supplier: TCI America D30715G
Specifications
| Diethyl (Bromodifluoromethyl)phosphonate | |
| Colorless | |
| C5H10BrF2O3P | |
| 5 g | |
| QRADKVYIJIAENZ-UHFFFAOYSA-N | |
| diethyl (bromodifluoromethyl)phosphonate | |
| 144101 | |
| ≥97.0% (GC) |
| 65094-22-6 | |
| 102°C | |
| MFCD00069118 | |
| diethyl bromodifluoromethyl phosphonate, bromodifluoromethyl diethylphosphonate, diethyl bromodifluoromethanephosphonate, diethyl bromodifluoromethylphosphonate, bromodifluoromethyl phosphonic acid diethyl ester, 1-bromo difluoro methyl-ethoxyphosphoryl oxyethane, phosphonic acid, bromodifluoromethyl-, diethyl ester, pubchem24183, acmc-20ai7p | |
| CCOP(=O)(OCC)C(F)(F)Br | |
| 267.01 | |
| 267.01 | |
| Liquid |
Chemical Identifiers
| 65094-22-6 | |
| 267.01 | |
| QRADKVYIJIAENZ-UHFFFAOYSA-N | |
| 144101 | |
| CCOP(=O)(OCC)C(F)(F)Br |
| C5H10BrF2O3P | |
| MFCD00069118 | |
| diethyl bromodifluoromethyl phosphonate, bromodifluoromethyl diethylphosphonate, diethyl bromodifluoromethanephosphonate, diethyl bromodifluoromethylphosphonate, bromodifluoromethyl phosphonic acid diethyl ester, 1-bromo difluoro methyl-ethoxyphosphoryl oxyethane, phosphonic acid, bromodifluoromethyl-, diethyl ester, pubchem24183, acmc-20ai7p | |
| diethyl (bromodifluoromethyl)phosphonate |
Safety and Handling
TSCA : No