missing translation for 'onlineSavingsMsg'
Learn More

Diethyl (Bromodifluoromethyl)phosphonate 97.0+%, TCI America™

Catalog No. p-7127603
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D30715G 5 g
D307125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D30715G Supplier TCI America Supplier No. D30715G

Chemical Identifiers

CAS 65094-22-6
Molecular Formula C5H10BrF2O3P
Molecular Weight (g/mol) 267.01
MDL Number MFCD00069118
InChI Key QRADKVYIJIAENZ-UHFFFAOYSA-N
Synonym diethyl bromodifluoromethyl phosphonate, bromodifluoromethyl diethylphosphonate, diethyl bromodifluoromethanephosphonate, diethyl bromodifluoromethylphosphonate, bromodifluoromethyl phosphonic acid diethyl ester, 1-bromo difluoro methyl-ethoxyphosphoryl oxyethane, phosphonic acid, bromodifluoromethyl-, diethyl ester, pubchem24183, acmc-20ai7p
PubChem CID 144101
IUPAC Name diethyl (bromodifluoromethyl)phosphonate
SMILES CCOP(=O)(OCC)C(F)(F)Br

Specifications

Color Colorless
Boiling Point 102°C
Quantity 5 g
Formula Weight 267.01
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl (Bromodifluoromethyl)phosphonate