missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl Benzylmalonate 98.0+%, TCI America™
Supplier: TCI America B0423500ML
Specifications
| Diethyl Benzylmalonate | |
| Colorless | |
| C14H18O4 | |
| 500 mL | |
| ICZLTZWATFXDLP-UHFFFAOYSA-N | |
| diethyl 2-benzylpropanedioate | |
| 69090 | |
| ≥98.0% (GC) |
| 607-81-8 | |
| 300°C | |
| MFCD00009166 | |
| diethyl benzylmalonate, diethyl 2-benzylmalonate, benzylmalonic acid diethyl ester, propanedioic acid, phenylmethyl-, diethyl ester, unii-40y0x0xn6p, malonic acid, benzyl-, diethyl ester, malonic acid, 2-benzyl-, diethyl ester, diethyl benzyl malonate, 1,3-diethyl 2-benzylpropanedioate, diethyl benzylpropanedioate | |
| CCOC(=O)C(CC1=CC=CC=C1)C(=O)OCC | |
| 250.294 | |
| 250.29 | |
| Liquid |
Chemical Identifiers
| 607-81-8 | |
| 250.294 | |
| ICZLTZWATFXDLP-UHFFFAOYSA-N | |
| 69090 | |
| CCOC(=O)C(CC1=CC=CC=C1)C(=O)OCC |
| C14H18O4 | |
| MFCD00009166 | |
| diethyl benzylmalonate, diethyl 2-benzylmalonate, benzylmalonic acid diethyl ester, propanedioic acid, phenylmethyl-, diethyl ester, unii-40y0x0xn6p, malonic acid, benzyl-, diethyl ester, malonic acid, 2-benzyl-, diethyl ester, diethyl benzyl malonate, 1,3-diethyl 2-benzylpropanedioate, diethyl benzylpropanedioate | |
| diethyl 2-benzylpropanedioate |
Safety and Handling
TSCA : Yes