missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Benzylmalonate 98.0+%, TCI America™

Catalog No. p-7118438
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B042325ML 25 mL
B0423500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B042325ML Supplier TCI America Supplier No. B042325ML
Only null left

Chemical Identifiers

CAS 607-81-8
Molecular Formula C14H18O4
Molecular Weight (g/mol) 250.294
MDL Number MFCD00009166
InChI Key ICZLTZWATFXDLP-UHFFFAOYSA-N
Synonym diethyl benzylmalonate, diethyl 2-benzylmalonate, benzylmalonic acid diethyl ester, propanedioic acid, phenylmethyl-, diethyl ester, unii-40y0x0xn6p, malonic acid, benzyl-, diethyl ester, malonic acid, 2-benzyl-, diethyl ester, diethyl benzyl malonate, 1,3-diethyl 2-benzylpropanedioate, diethyl benzylpropanedioate
PubChem CID 69090
IUPAC Name diethyl 2-benzylpropanedioate
SMILES CCOC(=O)C(CC1=CC=CC=C1)C(=O)OCC

Specifications

Color Colorless
Boiling Point 300°C
Quantity 25 mL
Formula Weight 250.29
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl Benzylmalonate