missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Benzylidenemalonate 96.0+%, TCI America™

Catalog No. p-7118436
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
B04205G 5 g
B042025G 25 g
2 options

Catalog No. B04205G

Supplier: TCI America B04205G

Only null left

Chemical Identifiers

CAS 5292-53-5
Molecular Formula C14H16O4
Molecular Weight (g/mol) 248.28
MDL Number MFCD00009149
InChI Key VUWPIBNKJSEYIN-UHFFFAOYSA-N
Synonym diethyl benzylidenemalonate, diethyl benzalmalonate, diethyl 2-benzylidenemalonate, ethyl benzylidenemalonate, diethylbenzalmalonate, diethyl phenylmethylene malonate, malonic acid, benzylidene-, diethyl ester, diethylmalonate, benzal, benzylidenemalonic acid diethyl ester, propanedioic acid, phenylmethylene-, diethyl ester
PubChem CID 94751
IUPAC Name 1,3-diethyl 2-(phenylmethylidene)propanedioate
SMILES CCOC(=O)C(=CC1=CC=CC=C1)C(=O)OCC

Specifications

Color Yellow
Quantity 5 g
Formula Weight 248.28
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl Benzylidenemalonate