accessibility menu, dialog, popup

UserName

CAS RN 5292-53-5

IUPAC Name: 1,3-diethyl 2-(phenylmethylidene)propanedioate
Synonyms: diethylmalonate, benzal
Molecular Weight (g/mol): 248.28
Molecular Formula: C14H16O4
InChi Key: VUWPIBNKJSEYIN-UHFFFAOYSA-N
SMILES: CCOC(=O)C(=CC1=CC=CC=C1)C(=O)OCC
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)