missing translation for 'onlineSavingsMsg'
Learn More

Diethyl benzylidenemalonate, 98%

Catalog No. p-7043392
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0056114 25 g
AAL0056122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0056114 Supplier Thermo Scientific Chemicals Supplier No. L0056114

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5292-53-5 | C14H16O4 | 248.28 g/mol

Diethyl benzylidenemalonate is used as a pharmaceutical intermediate. It is also involved in the Conjugate addition of cyanide ion to arylidenemalonic esters provides a useful route to arylsuccinic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl benzylidenemalonate is used as a pharmaceutical intermediate. It is also involved in the Conjugate addition of cyanide ion to arylidenemalonic esters provides a useful route to arylsuccinic acids.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.

Chemical Identifiers

CAS 5292-53-5
Molecular Formula C14H16O4
Molecular Weight (g/mol) 248.28
MDL Number MFCD00009149
InChI Key VUWPIBNKJSEYIN-UHFFFAOYSA-N
Synonym diethyl benzylidenemalonate, diethyl benzalmalonate, diethyl 2-benzylidenemalonate, ethyl benzylidenemalonate, diethylbenzalmalonate, diethyl phenylmethylene malonate, malonic acid, benzylidene-, diethyl ester, diethylmalonate, benzal, benzylidenemalonic acid diethyl ester, propanedioic acid, phenylmethylene-, diethyl ester
PubChem CID 94751
IUPAC Name diethyl 2-benzylidenepropanedioate
SMILES CCOC(=O)C(=CC1=CC=CC=C1)C(=O)OCC

Specifications

Melting Point 28°C to 31°C
Density 1.107
Boiling Point 215°C to 217°C (30 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.536
Quantity 25 g
Beilstein 528894
Solubility Information Insoluble in water.
Formula Weight 248.28
Percent Purity 98%
Chemical Name or Material Diethyl benzylidenemalonate
Show More Show Less
EINECSNumber 226-137-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only