missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Allylphosphonate 95.0+%, TCI America™

Catalog No. D30695G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D30695G 5 g
D30691G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D30695G Supplier TCI America Supplier No. D30695G

Chemical Identifiers

CAS 1067-87-4
Molecular Formula C7H15O3P
Molecular Weight (g/mol) 178.17
MDL Number MFCD00015134
InChI Key YPJHXRAHMUKXAE-UHFFFAOYSA-N
Synonym diethyl allylphosphonate, diethyl prop-2-en-1-ylphosphonate, diethyl-allylphosphonate, acmc-1bprn, phosphonic acid, 2-propenyl-, diethyl ester, allylphosphonic acid diethyl, 3-diethoxyphosphoryl-prop-1-ene, allylphosphonic acid diethyl ester, allyl phosphonic acid diethyl ester
PubChem CID 573014
IUPAC Name diethyl (prop-2-en-1-yl)phosphonate
SMILES CCOP(=O)(CC=C)OCC

Specifications

Color Colorless
Boiling Point 102°C
Quantity 5 g
Formula Weight 178.17
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl Allylphosphonate