accessibility menu, dialog, popup

UserName

CAS RN 1067-87-4

IUPAC Name: diethyl (prop-2-en-1-yl)phosphonate
Synonyms: diethyl prop-2-en-1-ylphosphonate
Molecular Weight (g/mol): 178.17
Molecular Formula: C7H15O3P
InChi Key: YPJHXRAHMUKXAE-UHFFFAOYSA-N
SMILES: CCOP(=O)(CC=C)OCC
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)