missing translation for 'onlineSavingsMsg'
Learn More

Diethyl allylphosphonate, 97%

Catalog No. p-108216
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC336110050 5 g Glass bottle
AC336110250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC336110050 Supplier Thermo Scientific Chemicals Supplier No. 336110050

CAS: 1067-87-4 | C7H15O3P | 178.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For C-C Bond Formation (Olefination)

Chemical Identifiers

CAS 1067-87-4
Molecular Formula C7H15O3P
Molecular Weight (g/mol) 178.17
MDL Number MFCD00015134
InChI Key YPJHXRAHMUKXAE-UHFFFAOYSA-N
Synonym diethyl allylphosphonate, diethyl prop-2-en-1-ylphosphonate, diethyl-allylphosphonate, acmc-1bprn, phosphonic acid, 2-propenyl-, diethyl ester, allylphosphonic acid diethyl, 3-diethoxyphosphoryl-prop-1-ene, allylphosphonic acid diethyl ester, allyl phosphonic acid diethyl ester
PubChem CID 573014
IUPAC Name 3-diethoxyphosphorylprop-1-ene
SMILES CCOP(=O)(CC=C)OCC

Specifications

Color Colorless
Boiling Point 97.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4325 to 1.4345
Linear Formula (O)P(CH3CH2O)2CH2CHCH2
Quantity 5 g
Formula Weight 178.17
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Diethyl allylphosphonate, 97%
Show More Show Less

RUO – Research Use Only