missing translation for 'onlineSavingsMsg'
Learn More

Diethyl adipate, ≥99.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111013071
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013071 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013071 Supplier Merck Emd Millipore Supplier No. 426971ML

Specifications

CAS 141-28-6
Density 1.009 g/mL (at 25°C)
Linear Formula C2H5 OCO(CH2)4 COOC2H5
MDL Number MFCD00009215
Melting Point -20°C to -19°C
Boiling Point 251°C
Synonym Hexanedioic acid 1,6-diethyl ester; NSC 19160; NSC 3363
Type Analytical Standard
Quantity 1 mL
Formula Weight 202.25
Percent Purity ≥99.0% (GC)
Grade Analytical Standard
Show More Show Less