missing translation for 'onlineSavingsMsg'
Learn More

Diethyl acetylsuccinate, 97+%

Catalog No. AAB2374406
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2374406 5 g
AAB2374414 25 g
AAB2374422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2374406 Supplier Thermo Scientific Chemicals Supplier No. B2374406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1115-30-6 | C10H16O5 | 216.233 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1115-30-6
Molecular Formula C10H16O5
Molecular Weight (g/mol) 216.233
MDL Number MFCD00009157
InChI Key DVSDDICSXBCMQJ-UHFFFAOYSA-N
Synonym diethyl 2-acetylsuccinate, diethyl acetylsuccinate, ethyl acetylsuccinate, diethyl acetosuccinate, butanedioic acid, acetyl-, diethyl ester, acetylsuccinic acid diethyl ester, diethylacetylsuccinate, 1,4-diethyl 2-acetylbutanedioate, succinic acid, acetyl-, diethyl ester, butanedioic acid, 2-acetyl-, 1,4-diethyl ester
PubChem CID 66197
IUPAC Name diethyl 2-acetylbutanedioate
SMILES CCOC(=O)CC(C(=O)C)C(=O)OCC

Specifications

Melting Point -8°C
Density 1.081
Boiling Point 180°C to 183°C (50 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.4346
Quantity 5 g
Formula Weight 216.23
Percent Purity ≥97%
Chemical Name or Material Diethyl acetylsuccinate
EINECSNumber 214-223-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only