accessibility menu, dialog, popup

UserName

CAS RN 1115-30-6

IUPAC Name: 1,4-diethyl 2-acetylbutanedioate
Synonyms: 1,4-diethyl 2-acetylbutanedioate
Molecular Weight (g/mol): 216.23
Molecular Formula: C10H16O5
InChi Key: DVSDDICSXBCMQJ-UHFFFAOYNA-N
SMILES: CCOC(=O)CC(C(C)=O)C(=O)OCC
Boiling Point 
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (6)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)