missing translation for 'onlineSavingsMsg'
Learn More

Diethyl acetylmethylphosphonate, 97%, Thermo Scientific Chemicals

Catalog No. AAH2689606
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2689606 5 g
AAH2689614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2689606 Supplier Thermo Scientific Chemicals Supplier No. H2689606

Diethyl acetylmethylphosphonate is used as ligand in coordination chemistry to prepare lanthanide series metals like terbium, europium and neodymium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl acetylmethylphosphonate is used as ligand in coordination chemistry to prepare lanthanide series metals like terbium, europium and neodymium.

Solubility
Miscible with tetrahydrofuran, ether, dichloromethane and chloroform.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 1067-71-6
Molecular Formula C7H15O4P
Molecular Weight (g/mol) 194.167
MDL Number MFCD00044728
InChI Key RSAFKRSMGOSHRK-UHFFFAOYSA-N
Synonym diethyl 2-oxopropyl phosphonate, diethyl acetonylphosphonate, diethyl 2-oxopropylphosphonate, acetonylphosphonic acid diethyl ester, 2-oxopropyl phosphonic acid diethyl ester, phosphonic acid, 2-oxopropyl-, diethyl ester, aurora ka-1476, ksc490a4t, diethyl acetylmethylphosphonate
PubChem CID 14028
IUPAC Name 1-diethoxyphosphorylpropan-2-one
SMILES CCOP(=O)(CC(=O)C)OCC

Specifications

Density 1.01
Boiling Point 129°C (9 mmHg)
Flash Point 113°C (235°F)
Refractive Index 1.433
Quantity 5 g
Solubility Information Miscible with tetrahydrofuran,ether,dichloromethane and chloroform.
Formula Weight 194.17
Percent Purity 97%
Chemical Name or Material Diethyl acetylmethylphosphonate
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only