missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 4-Methoxyphenylphosphonate 97.0+%, TCI America™

Catalog No. M115210G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M115210G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M115210G Supplier TCI America Supplier No. M115210G
Only null left

Chemical Identifiers

CAS 3762-33-2
Molecular Formula C11H17O4P
Molecular Weight (g/mol) 244.227
MDL Number MFCD00015670
InChI Key OTUVLLBGFFIEJO-UHFFFAOYSA-N
Synonym 4-Methoxyphenylphosphonic Acid Diethyl Ester
PubChem CID 604767
IUPAC Name 1-diethoxyphosphoryl-4-methoxybenzene
SMILES CCOP(=O)(C1=CC=C(C=C1)OC)OCC

Specifications

Color Colorless
Quantity 10 g
Formula Weight 244.23
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl 4-Methoxyphenylphosphonate