accessibility menu, dialog, popup

UserName

CH3OPOOCH3OCH3

CAS RN 3762-33-2

IUPAC Name: diethyl (4-methoxyphenyl)phosphonate
Synonyms: diethyl 4-methoxyphenylphosphonate
Molecular Weight (g/mol): 244.23
Molecular Formula: C11H17O4P
InChi Key: OTUVLLBGFFIEJO-UHFFFAOYSA-N
SMILES: CCOP(=O)(OCC)C1=CC=C(OC)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results