missing translation for 'onlineSavingsMsg'
Learn More

Diethyl (4-Cyanobenzyl)phosphonate 98.0+%, TCI America™

Catalog No. D33235G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D33235G 5 g
D332325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D33235G Supplier TCI America Supplier No. D33235G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1552-41-6
Molecular Formula C12H16NO3P
Molecular Weight (g/mol) 253.24
MDL Number MFCD08274488
InChI Key MFXWOYIWKNJHPC-UHFFFAOYSA-N
Synonym (4-Cyanobenzyl)phosphonic Acid Diethyl Ester, 4-(Diethylphosphonomethyl)benzonitrile
PubChem CID 241438
IUPAC Name diethyl [(4-cyanophenyl)methyl]phosphonate
SMILES CCOP(=O)(CC1=CC=C(C=C1)C#N)OCC

Specifications

Melting Point 31°C
Color White-Yellow
Boiling Point 169°C
Quantity 5 g
UN Number 3439
Formula Weight 253.24
Percent Purity ≥98.0% (GC)
Physical Form Crystals
Chemical Name or Material Diethyl (4-Cyanobenzyl)phosphonate