accessibility menu, dialog, popup

UserName

H3COPONOCH3

CAS RN 1552-41-6

IUPAC Name: diethyl [(4-cyanophenyl)methyl]phosphonate
Synonyms: diethyl (4-cyanophenyl)methylphosphonate
Molecular Weight (g/mol): 253.24
Molecular Formula: C12H16NO3P
InChi Key: MFXWOYIWKNJHPC-UHFFFAOYSA-N
SMILES: CCOP(=O)(CC1=CC=C(C=C1)C#N)OCC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results