missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 4,4-difluoropimelate, 97%, Thermo Scientific™

Catalog No. p-7030712
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3154403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3154403 Supplier Thermo Scientific Chemicals Supplier No. H3154403

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 22515-16-8
Molecular Formula C11H18F2O4
Molecular Weight (g/mol) 252.26
MDL Number MFCD08146634
InChI Key XUOBBVMKXUPPEW-UHFFFAOYSA-N
Synonym 4,4-difluoroheptanedioic acid diethyl ester, 1,7-diethyl 4,4-difluoroheptanedioate, diethyl 4,4-difluoropimelate, pubchem22650, diethyl 4,4-heptanedioate, ksc494q0l, diethyl4,4-difluoroheptanedioate, diethyl 4,4-difluoroheptane-1,7-dioate, 4,4-difluoro-heptanedioic acid diethyl ester, heptanedioicacid,4,4-difluoro-,1,7-diethylester
PubChem CID 10264195
IUPAC Name diethyl 4,4-difluoroheptanedioate
SMILES CCOC(=O)CCC(F)(F)CCC(=O)OCC

Specifications

Boiling Point 108°C (3 mmHg)
Refractive Index 1.415
Quantity 1 g
Formula Weight 252.26
Percent Purity 97%
Chemical Name or Material Diethyl 4,4-difluoropimelate
TSCA No
Recommended Storage Ambient temperatures