missing translation for 'onlineSavingsMsg'
Learn More

Diethyl (3-Methoxybenzyl)phosphonate 98.0+%, TCI America™

Catalog No. D33265G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D33265G 5 g
D332625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D33265G Supplier TCI America Supplier No. D33265G
Only null left

Chemical Identifiers

CAS 60815-18-1
Molecular Formula C12H19O4P
Molecular Weight (g/mol) 258.25
MDL Number MFCD08276363
InChI Key IGGMUQNGGBRXFB-UHFFFAOYSA-N
Synonym 3-(Diethylphosphonomethyl)anisole, (3-Methoxybenzyl)phosphonic Acid Diethyl Ester
PubChem CID 10611200
IUPAC Name diethyl [(3-methoxyphenyl)methyl]phosphonate
SMILES CCOP(=O)(CC1=CC(OC)=CC=C1)OCC

Specifications

Color Yellow
Boiling Point 135°C
Quantity 5 g
Formula Weight 258.25
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl (3-Methoxybenzyl)phosphonate