missing translation for 'onlineSavingsMsg'
Learn More

Diethyl (3-chloropropyl)malonate, 98%

Catalog No. AAL1477614
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1477614 25 g
AAL1477606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1477614 Supplier Thermo Scientific Chemicals Supplier No. L1477614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 18719-43-2 | C10H17ClO4 | 236.692 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 18719-43-2
Molecular Formula C10H17ClO4
Molecular Weight (g/mol) 236.692
MDL Number MFCD00009169
InChI Key WUOPYKKJFZHTRA-UHFFFAOYSA-N
Synonym diethyl 3-chloropropyl malonate, diethyl 2-3-chloropropyl malonate, malonic acid, 3-chloropropyl-, diethyl ester, 2-3-chloropropyl malonic acid diethyl ester, acmc-20al9u, diethyl ?-chloropropyl malonate, chloropropylmalonic acid diethyl ester, diethyl 2-3-chloropropyl propanedioate
PubChem CID 140413
IUPAC Name diethyl 2-(3-chloropropyl)propanedioate
SMILES CCOC(=O)C(CCCCl)C(=O)OCC

Specifications

Density 1.101
Boiling Point 102°C (1 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.443
Quantity 25 g
Beilstein 1785711
Formula Weight 236.7
Percent Purity 98%
Chemical Name or Material Diethyl (3-chloropropyl)malonate
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only