accessibility menu, dialog, popup

UserName

CH3OOClOOCH3

CAS RN 18719-43-2

IUPAC Name: 1,3-diethyl 2-(3-chloropropyl)propanedioate
Synonyms: 1,3-diethyl 2-(3-chloropropyl)propanedioate
Molecular Weight (g/mol): 236.69
Molecular Formula: C10H17ClO4
InChi Key: WUOPYKKJFZHTRA-UHFFFAOYSA-N
SMILES: CCOC(=O)C(CCCCl)C(=O)OCC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results