missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl 3,5-Dimethyl-2,4-pyrroledicarboxylate 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 136°C |
| Quantity | 5 g |
| Synonym | diethyl 2,4-dimethylpyrrole-3,5-dicarboxylate, 3,5-dimethyl-1h-pyrrole-2,4-dicarboxylic acid diethyl ester, knorr's pyrrole, 2,4-diethyl 3,5-dimethyl-1h-pyrrole-2,4-dicarboxylate, 1h-pyrrole-2,4-dicarboxylic acid, 3,5-dimethyl-, diethyl ester, 2,4-dimethyl-3,5-dicarbethoxypyrrole, diethyl 2,4-dimethyl-1h-pyrrole-3,5-dicarboxylate, diethyl2,4-dimethylpyrrole-3,5-dicarboxylate, pyrrole-2,4-dicarboxylic acid, 3,5-dimethyl-, diethyl ester, diethyl 2 ,4-dimethylpyrrole-3,5-dicarboxylate |
| Formula Weight | 239.27 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Diethyl 3,5-Dimethyl-2,4-pyrroledicarboxylate |
Safety and Handling
| TSCA | No |