missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 3,5-Di-tert-butyl-4-hydroxybenzylphosphonate 98.0+%, TCI America™

Catalog No. D29671G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D29671G 1 g
D29675G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D29671G Supplier TCI America Supplier No. D29671G
Only null left

Chemical Identifiers

CAS 976-56-7
Molecular Formula C19H33O4P
Molecular Weight (g/mol) 356.44
MDL Number MFCD00227871
InChI Key GJDRKHHGPHLVNI-UHFFFAOYSA-N
Synonym 3,5-Di-tert-butyl-4-hydroxybenzylphosphonic Acid Diethyl Ester
PubChem CID 70421
IUPAC Name diethyl [(3,5-di-tert-butyl-4-hydroxyphenyl)methyl]phosphonate
SMILES CCOP(=O)(CC1=CC(=C(O)C(=C1)C(C)(C)C)C(C)(C)C)OCC

Specifications

Melting Point 122°C
Color White
Quantity 1 g
Formula Weight 356.44
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Diethyl 3,5-Di-tert-butyl-4-hydroxybenzylphosphonate
EINECSNumber (3)-3376
TSCA Yes