missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 3,4-furandicarboxylate, 95%

Catalog No. ac156510050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC156510050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC156510050 Supplier Thermo Scientific Chemicals Supplier No. 156510050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 30614-77-8 | C10H12O5 | 212.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 30614-77-8
Molecular Formula C10H12O5
Molecular Weight (g/mol) 212.20
MDL Number MFCD00005349
InChI Key ZODFWNHYQARJLC-UHFFFAOYSA-N
Synonym diethyl 3,4-furandicarboxylate, 3,4-furandicarboxylic acid, diethyl ester, 3,4-diethyl furan-3,4-dicarboxylate, furan-3,4-dicarboxylic acid diethyl ester, 5-18-07-00053 beilstein handbook reference, zodfwnhyqarjlc-uhfffaoysa, 3,4-furandicarboxylicacid, 3,4-diethyl ester
PubChem CID 35399
IUPAC Name diethyl furan-3,4-dicarboxylate
SMILES CCOC(=O)C1=COC=C1C(=O)OCC

Specifications

Color Colorless
Density 1.1400g/mL
Boiling Point 155°C (13.0 mmHg)
Flash Point 82°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4715 to 1.4735
Quantity 5 g
Specific Gravity 1.14
Formula Weight 212.2
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Diethyl 3, 4-furandicarboxylate
Show More Show Less
Hazard Statement GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 250-262-7

RUO – Research Use Only