accessibility menu, dialog, popup

UserName

CAS RN 30614-77-8

IUPAC Name: 3,4-diethyl furan-3,4-dicarboxylate
Synonyms: 3,4-diethyl furan-3,4-dicarboxylate
Molecular Weight (g/mol): 212.2
Molecular Formula: C10H12O5
InChi Key: ZODFWNHYQARJLC-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=COC=C1C(=O)OCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results