missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 3,3'-Iminodipropionate 98.0+%, TCI America™

Catalog No. D41115G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D41115G 5 g
D41111G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D41115G Supplier TCI America Supplier No. D41115G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3518-88-5
Molecular Formula C10H19NO4
Molecular Weight (g/mol) 217.265
MDL Number MFCD14705114
InChI Key ONEIYJQBXMVMKU-UHFFFAOYSA-N
PubChem CID 352310
IUPAC Name ethyl 3-[(3-ethoxy-3-oxopropyl)amino]propanoate
SMILES CCOC(=O)CCNCCC(=O)OCC

Specifications

Boiling Point 139°C
Color Colorless
Synonym 3,3′-Iminodipropionic Acid Diethyl Ester
Quantity 5 g
Formula Weight 217.27
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Diethyl 3,3′-Iminodipropionate
TSCA No
Recommended Storage Refrigerator